siRNA Details
| Gene Symbol: SULT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003167.3 | NP_003158.2 | 5 | 770 | ||
| GenBank mRNA | AK289380.1 | - | 5 | 712 | ||
| AK313415.1 | - | 5 | 697 | |||
| BC020755.1 | - | 5 | 687 | 4733628 | ||
| L02337.1 | - | 5 | 695 | |||
| L20000.1 | - | 5 | 682 | |||
| S43859.1 | - | 5 | 695 | |||
| S43861.1 | - | 4 | 464 | |||
| U08024.1 | - | 5 | 650 | |||
| U08025.1 | - | 5 | 694 | |||
| X70222.1 | - | 5 | 682 | |||
| X84816.1 | - | 5 | 675 | |||

SULT2A1
Genomic Map