siRNA Details
| Gene Symbol: TST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270483.1 | NP_001257412.1 | 2 | 419 | ||
| NM_003312.5 | NP_003303.2 | 2 | 451 | |||
| GenBank mRNA | AK000862.1 | - | 2 | 445 | ||
| AK091874.1 | - | 1 | 1071 | |||
| AK223521.1 | - | 2 | 445 | |||
| BC010148.2 | - | 2 | 428 | 4329727 | ||
| BI820468.1 | - | 2 | 448 | |||
| BQ231617.1 | - | 2 | 444 | |||
| CR456598.1 | - | 1 | 454 | |||
| CR456998.1 | - | 1 | 376 | |||
| D87292.1 | - | 2 | 424 | |||

TST
Genomic Map