siRNA Details
| Gene Symbol: TST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270483.1 | NP_001257412.1 | 3 | 1004 | ||
| NM_003312.5 | NP_003303.2 | 3 | 1036 | |||
| GenBank mRNA | AK000862.1 | - | 3 | 1030 | ||
| AK091874.1 | - | 2 | 1656 | |||
| AK223521.1 | - | 3 | 1030 | |||
| AK301300.1 | - | 4 | 871 | |||
| BC010148.2 | - | 3 | 1013 | 4329727 | ||
| CR456598.1 | - | 2 | 1039 | |||
| D87292.1 | - | 3 | 1009 | |||

TST
Genomic Map