siRNA Details
| Gene Symbol: KRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002275.3 | NP_002266.2 | 1 | 314 | ||
| XM_011524784.2 | XP_011523086.1 | 2 | 1897 | |||
| XM_017024614.1 | XP_016880103.1 | 1 | 360 | |||
| GenBank mRNA | AK090604.1 | - | 3 | 867 | ||
| AK122864.1 | - | 1 | 3709 | |||
| AK304059.1 | - | 1 | 186 | |||
| AL832226.1 | - | 1 | 2315 | |||
| BC002641.2 | - | 1 | 179 | 3610392 | ||
| BT007261.1 | - | 1 | 123 | |||
| X07696.1 | - | 1 | 184 | |||

KRT15
Genomic Map