siRNA Details
| Gene Symbol: B4GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005417.2 | NP_001005417.1 | 7 | 1661 | ||
| NM_003780.4 | NP_003771.1 | 7 | 1861 | |||
| NM_030587.2 | NP_085076.2 | 7 | 1662 | |||
| XM_017002716.1 | XP_016858205.1 | 7 | 1619 | |||
| XM_017002717.1 | XP_016858206.1 | 7 | 1822 | |||
| GenBank mRNA | AF038660.1 | - | 6 | 1549 | ||
| AK074533.1 | - | 5 | 1159 | |||
| AK095873.1 | - | 7 | 2289 | |||
| AL137647.1 | - | 5 | 1056 | |||
| BC096821.1 | - | 7 | 1629 | 30558762 | ||

B4GALT2
Genomic Map