siRNA Details
| Gene Symbol: LAG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002286.5 | NP_002277.4 | 6 | 1467 | ||
| GenBank mRNA | A21353.1 | - | 6 | 1348 | ||
| AK292104.1 | - | 6 | 1441 | |||
| X51985.3 | - | 6 | 1348 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAG3| Gene Symbol: LAG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002286.5 | NP_002277.4 | 6 | 1467 | ||
| GenBank mRNA | A21353.1 | - | 6 | 1348 | ||
| AK292104.1 | - | 6 | 1441 | |||
| X51985.3 | - | 6 | 1348 | |||