siRNA Details
| Gene Symbol: LARGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004737.4 | NP_004728.1 | 16 | 3137 | ||
| NM_133642.3 | NP_598397.1 | 15 | 3074 | |||
| XM_005261831.3 | XP_005261888.1 | 16 | 2983 | |||
| XM_005261832.3 | XP_005261889.1 | 15 | 2938 | |||
| XM_011530510.2 | XP_011528812.1 | 16 | 2924 | |||
| XM_011530512.2 | XP_011528814.1 | 11 | 2061 | |||
| XM_011530513.2 | XP_011528815.1 | 7 | 1602 | |||
| GenBank mRNA | AB011181.3 | - | 14 | 2833 | ||
| AJ007583.1 | - | 16 | 3047 | |||
| BC117425.1 | - | 14 | 2626 | 40125976 | ||
| BC126404.1 | - | 14 | 2626 | 8992120 | ||

LARGE1
Genomic Map