siRNA Details
| Gene Symbol: SHMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166356.1 | NP_001159828.1 | 8 | 1120 | ||
| NM_001166357.1 | NP_001159829.1 | 8 | 1212 | |||
| NM_001166358.1 | NP_001159830.1 | 8 | 1111 | |||
| NM_001166359.1 | NP_001159831.1 | 8 | 1004 | |||
| NM_005412.5 | NP_005403.2 | 8 | 1150 | |||
| NR_029415.1 | 7 | 952 | ||||
| NR_029416.1 | 7 | 1031 | ||||
| NR_029417.1 | 7 | 1070 | ||||
| NR_048562.1 | 7 | 1160 | ||||
| XM_011538675.2 | XP_011536977.1 | 8 | 1090 | |||
| XM_011538676.2 | XP_011536978.1 | 8 | 1081 | |||
| XM_011538677.2 | XP_011536979.1 | 7 | 991 | |||
| XM_011538678.2 | XP_011536980.1 | 7 | 973 | |||
| GenBank mRNA | AK055053.1 | - | 4 | 2136 | ||
| AK223555.1 | - | 8 | 1012 | |||
| AK295941.1 | - | 7 | 1006 | |||
| AK296183.1 | - | 8 | 964 | |||
| AK297173.1 | - | 7 | 912 | |||
| AK298231.1 | - | 7 | 805 | |||
| AK301339.1 | - | 8 | 877 | |||
| AK301443.1 | - | 7 | 974 | |||
| AK303827.1 | - | 7 | 826 | |||
| AK315916.1 | - | 8 | 1204 | |||
| BC011911.2 | - | 8 | 954 | 4139001 | ||
| BC013677.2 | - | 8 | 1014 | 3863368 | ||
| BC025355.1 | - | 5 | 537 | |||
| BC032584.1 | - | 8 | 993 | 5499481 | ||
| BC044211.1 | - | 8 | 990 | 5521082 | ||
| BC091501.1 | - | 7 | 874 | |||
| BT006866.1 | - | 8 | 944 | |||
| BX647711.1 | - | 8 | 1071 | |||
| BX648847.1 | - | 6 | 552 | |||
| EU668344.1 | - | 8 | 954 | |||
| KU178333.1 | - | 8 | 944 | |||
| KU178334.1 | - | 7 | 580 | |||
| L11932.1 | - | 7 | 855 | |||

SHMT2
Genomic Map