siRNA Details
| Gene Symbol: SULT1E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005420.2 | NP_005411.1 | 5, 6 | 616 | ||
| GenBank mRNA | AK312236.1 | - | 4, 5 | 515 | ||
| BC027956.1 | - | 5, 6 | 616 | 5210178 | ||
| CR407621.1 | - | 4, 5 | 503 | |||
| S77383.1 | - | 5, 6 | 539 | |||
| U08098.1 | - | 5, 6 | 609 | |||
| Y11195.1 | - | 5, 6 | 565 | |||

SULT1E1
Genomic Map