siRNA Details
| Gene Symbol: CEPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007794.1 | NP_001007795.1 | 2 | 354 | ||
| NM_006090.3 | NP_006081.1 | 2 | 316 | |||
| XM_006710285.3 | XP_006710348.1 | 2 | 308 | |||
| XM_011540496.1 | XP_011538798.1 | 2 | 316 | |||
| GenBank mRNA | AF068302.1 | - | 2 | 274 | ||
| AF138862.1 | - | 1 | 184 | |||
| AK023381.1 | - | 2 | 303 | |||
| AL833102.1 | - | 2 | 327 | |||
| BC032610.2 | - | 3 | 316 | 5554219 | ||
| BC049196.1 | - | 2 | 354 | 5226538 | ||
| BX487092.1 | - | 2 | 316 | |||

CEPT1
Genomic Map