siRNA Details
| Gene Symbol: ID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002165.3 | NP_002156.2 | 1 | 377 | ||
| NM_181353.2 | NP_851998.1 | 1 | 377 | |||
| GenBank mRNA | AK291152.1 | - | 1 | 295 | ||
| AK313786.1 | - | 1 | 372 | |||
| BC000613.2 | - | 1 | 338 | 3346009 | ||
| BC012420.1 | - | 1 | 358 | 3862019 | ||
| BG546814.1 | - | 1 | 372 | |||
| BI768231.1 | - | 1 | 224 | |||
| BT007443.1 | - | 1 | 272 | |||
| D13889.1 | - | 1 | 290 | |||
| D13890.1 | - | 1 | 290 | |||
| DA839689.1 | - | 1 | 377 | |||
| S78825.1 | - | 1 | 282 | |||
| S78986.1 | - | 1 | 334 | |||
| X77956.1 | - | 1 | 304 | |||

ID1
Genomic Map