siRNA Details
| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321770.1 | NP_001308699.1 | 1 | 504 | ||
| NM_006588.3 | NP_006579.2 | 1 | 504 | |||
| NR_135776.1 | 1 | 504 | ||||
| NR_135779.1 | 1 | 504 | ||||
| GenBank mRNA | AK297851.1 | - | 1 | 282 | ||
| AK298755.1 | - | 1 | 445 | |||
| BC058861.1 | - | 1 | 504 | |||
| BC125043.1 | - | 1 | 230 | 40115974 | ||

SULT1C4
Genomic Map