siRNA Details
| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006588.3 | NP_006579.2 | 3 | 742 | ||
| XM_017003807.1 | XP_016859296.1 | 3 | 284 | |||
| GenBank mRNA | BC058861.1 | - | 2 | 1239 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SULT1C4| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006588.3 | NP_006579.2 | 3 | 742 | ||
| XM_017003807.1 | XP_016859296.1 | 3 | 284 | |||
| GenBank mRNA | BC058861.1 | - | 2 | 1239 | ||