siRNA Details
| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321770.1 | NP_001308699.1 | 5 | 1328 | ||
| NM_006588.3 | NP_006579.2 | 7 | 1553 | |||
| NR_135776.1 | 5 | 1378 | ||||
| NR_135779.1 | 3 | 1107 | ||||
| XM_017003807.1 | XP_016859296.1 | 7 | 1095 | |||
| GenBank mRNA | AK298755.1 | - | 6 | 917 | ||

SULT1C4
Genomic Map