siRNA Details
| Gene Symbol: INMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199219.1 | NP_001186148.1 | 3 | 645 | ||
| NM_006774.4 | NP_006765.4 | 3 | 648 | |||
| GenBank mRNA | AF128846.1 | - | 3 | 648 | ||
| AF128847.1 | - | 3 | 648 | |||
| AK309519.1 | - | 4 | 778 | |||
| AK313832.1 | - | 3 | 643 | |||
| BC033813.1 | - | 3 | 630 | |||
| BC103712.1 | - | 3 | 631 | |||
| BC106902.1 | - | 3 | 648 | 40031569 | ||
| BC106903.1 | - | 3 | 648 | 40031570 | ||

INMT
Genomic Map