siRNA Details
| Gene Symbol: IDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289910.1 | NP_001276839.1 | 5 | 479 | ||
| NM_001290114.1 | NP_001277043.1 | 3 | 454 | |||
| NM_002168.3 | NP_002159.2 | 5 | 712 | |||
| GenBank mRNA | AK222916.1 | - | 5 | 626 | ||
| AK294148.1 | - | 5 | 479 | |||
| AK299987.1 | - | 3 | 368 | |||
| AK312627.1 | - | 5 | 676 | |||
| AK316388.1 | - | 5 | 553 | |||
| BC009244.2 | - | 5 | 614 | 2959540 | ||
| BC071828.1 | - | 5 | 613 | 6090792 | ||
| X69433.1 | - | 5 | 634 | |||

IDH2
Genomic Map