siRNA Details
| Gene Symbol: LILRB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081442.2 | NP_001074911.1 | 4 | 722 | ||
| NM_001304457.1 | NP_001291386.1 | 4 | 695 | |||
| NM_006840.4 | NP_006831.1 | 4 | 722 | |||
| XM_011526359.2 | XP_011524661.1 | 4 | 706 | |||
| XM_011526360.2 | XP_011524662.1 | 4 | 706 | |||
| XM_011526361.2 | XP_011524663.1 | 4 | 706 | |||
| GenBank mRNA | AF025534.1 | - | 4 | 673 | ||
| AK092867.1 | - | 4 | 590 | |||
| AK223296.1 | - | 4 | 678 | |||
| AK290845.1 | - | 4 | 706 | |||
| AK301112.1 | - | 4 | 658 | |||
| AK301338.1 | - | 3, 4 | 455 | |||
| BC025704.1 | - | 4 | 676 | 5223581 | ||

LILRB5
Genomic Map