siRNA Details
| Gene Symbol: IFNA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002172.2 | NP_002163.2 | 1 | 159 | ||
| GenBank mRNA | BC074956.2 | - | 1 | 159 | 30915281 | |
| BC104159.1 | - | 1 | 157 | 40029547 | ||
| BC104160.1 | - | 1 | 157 | 40029549 | ||
| V00542.1 | - | 1 | 171 | |||

IFNA14
Genomic Map