siRNA Details
| Gene Symbol: OGFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007346.2 | NP_031372.2 | 2 | 249 | ||
| GenBank mRNA | AF109134.1 | - | 2 | 242 | ||
| AF172449.1 | - | 2 | 247 | |||
| AF172450.1 | - | 2 | 247 | |||
| AF172451.1 | - | 2 | 247 | |||
| AF172452.1 | - | 2 | 247 | |||
| AF172453.1 | - | 247 | ||||
| AK022234.1 | - | 2 | 239 | |||
| AK024485.1 | - | 1 | 1694 | |||
| AK314696.1 | - | 2 | 241 | |||
| BC008768.2 | - | 2 | 224 | 3050950 | ||
| BC014137.2 | - | 2 | 224 | 4643904 | ||
| BC032666.1 | - | 2 | 204 | 5552531 | ||
| BM551600.1 | - | 2 | 249 | |||

OGFR
Genomic Map