siRNA Details
| Gene Symbol: POMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011536675.2 | XP_011534977.1 | 13 | 1580 | ||
| XM_011536676.2 | XP_011534978.1 | 12 | 1493 | |||
| XM_011536677.2 | XP_011534979.1 | 9 | 1122 | |||
| XM_011536679.2 | XP_011534981.1 | 9 | 782 | |||
| XM_017021244.1 | XP_016876733.1 | 12 | 1414 | |||
| XM_017021245.1 | XP_016876734.1 | 9 | 868 | |||
| GenBank mRNA | AF105020.1 | - | 13 | 1583 | ||
| AY090480.1 | - | 13 | 1583 | |||

POMT2
Genomic Map