siRNA Details
| Gene Symbol: POMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013382.5 | NP_037514.2 | 21 | 4025 | ||
| XM_011536675.2 | XP_011534977.1 | 22 | 4214 | |||
| XM_011536676.2 | XP_011534978.1 | 21 | 4127 | |||
| XM_011536677.2 | XP_011534979.1 | 18 | 3756 | |||
| XM_011536679.2 | XP_011534981.1 | 18 | 3416 | |||
| XM_017021244.1 | XP_016876733.1 | 20 | 3859 | |||
| XM_017021245.1 | XP_016876734.1 | 17 | 3313 | |||
| GenBank mRNA | AB209114.1 | - | 20 | 4800 | ||
| AK023804.1 | - | 15 | 4616 | |||
| AL353956.1 | - | 4 | 1991 | |||
| AY090480.1 | - | 21 | 4027 | |||
| BM724049.1 | - | 1 | 245 | |||

POMT2
Genomic Map