siRNA Details
| Gene Symbol: ST6GALNAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175039.3 | NP_778204.1 | 6 | 1673 | ||
| NM_175040.3 | NP_778205.1 | 5 | 1586 | |||
| XM_011518527.2 | XP_011516829.1 | 7 | 1765 | |||
| XM_011518529.2 | XP_011516831.1 | 7 | 1796 | |||
| GenBank mRNA | AK292049.1 | - | 6 | 1428 | ||
| BC036705.2 | - | 6 | 1576 | 4309208 | ||

ST6GALNAC4
Genomic Map