siRNA Details
| Gene Symbol: NDOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144026.2 | NP_001137498.1 | 6 | 753 | ||
| NM_001144027.2 | NP_001137499.1 | 5 | 651 | |||
| NM_001144028.2 | NP_001137500.1 | 6 | 753 | |||
| NM_014434.3 | NP_055249.1 | 6 | 753 | |||
| XM_011518545.1 | XP_011516847.1 | 5 | 641 | |||
| XM_011518547.1 | XP_011516849.1 | 6 | 543 | |||
| XM_011518548.2 | XP_011516850.1 | 5 | 1912 | |||
| XM_017014626.1 | XP_016870115.1 | 6 | 753 | |||
| XM_017014627.1 | XP_016870116.1 | 6 | 753 | |||
| GenBank mRNA | AF199509.1 | - | 6 | 689 | ||
| AK026089.1 | - | 6 | 725 | |||
| AK074403.1 | - | 5 | 612 | |||
| AK225995.1 | - | 6 | 724 | |||
| AK290026.1 | - | 6 | 750 | |||
| AY077845.1 | - | 6 | 671 | |||
| BC015735.1 | - | 6 | 689 | 4869561 | ||
| BC078668.1 | - | 6 | 689 | |||
| BC093782.1 | - | 6 | 670 | 7939627 | ||
| BC111943.1 | - | 6 | 670 | 8327411 | ||

NDOR1
Genomic Map