siRNA Details
| Gene Symbol: MICAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282663.1 | NP_001269592.1 | 28 | 3802 | ||
| NM_001282664.1 | NP_001269593.1 | 26 | 3592 | |||
| NM_001282665.1 | NP_001269594.1 | 22 | 4107 | |||
| NM_001282666.1 | NP_001269595.1 | 19 | 6506 | |||
| NM_001282667.1 | NP_001269596.1 | 18 | 6443 | |||
| NM_014632.3 | NP_055447.1 | 28 | 3806 | |||
| XM_017018589.1 | XP_016874078.1 | 26 | 4644 | |||
| XM_017018590.1 | XP_016874079.1 | 27 | 3667 | |||
| XM_017018591.1 | XP_016874080.1 | 25 | 4581 | |||
| GenBank mRNA | AB018293.1 | - | 28 | 3767 | ||
| AF052170.1 | - | 11 | 1383 | |||
| AK025186.1 | - | 1 | 2068 | |||
| AK026116.1 | - | 1 | 2228 | |||
| AK090550.1 | - | 1 | 3638 | |||
| AK122821.1 | - | 6 | 3869 | |||
| AK125552.1 | - | 1 | 3000 | |||
| AK125785.1 | - | 1 | 4419 | |||
| AK127301.1 | - | 4 | 5121 | |||
| AL046982.1 | - | 1 | 357 | |||
| BC017104.1 | - | 1 | 1939 | |||
| BC044577.1 | - | 28 | 3791 | 5275364 | ||
| BC048206.1 | - | 1 | 1863 | |||
| BX538021.1 | - | 16 | 6193 | |||
| BX641163.1 | - | 4 | 4233 | |||

MICAL2
Genomic Map