siRNA Details
| Gene Symbol: INSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002196.2 | NP_002187.1 | 1 | 1954 | ||
| GenBank mRNA | M93119.1 | - | 1 | 1954 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

INSM1| Gene Symbol: INSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002196.2 | NP_002187.1 | 1 | 1954 | ||
| GenBank mRNA | M93119.1 | - | 1 | 1954 | ||