siRNA Details
| Gene Symbol: ST8SIA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015879.2 | NP_056963.2 | 1 | 451 | ||
| GenBank mRNA | AB208996.1 | - | 1 | 455 | ||
| AF003092.2 | - | 1 | 166 | |||
| AF004668.2 | - | 1 | 2117 | |||
| AK289517.1 | - | 1 | 229 | |||
| AK313047.1 | - | 1 | 177 | |||
| BC074909.2 | - | 1 | 142 | 30915256 | ||

ST8SIA3
Genomic Map