siRNA Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291822.1 | NP_001278751.1 | 5 | 833 | ||
| NM_001291823.1 | NP_001278752.1 | 3 | 580 | |||
| NM_016523.2 | NP_057607.1 | 6 | 983 | |||
| NR_120305.1 | 4 | 693 | ||||
| XM_017019414.1 | XP_016874903.1 | 4 | 849 | |||
| GenBank mRNA | AF175206.1 | - | 6 | 983 | ||
| AF175207.1 | - | 5 | 833 | |||
| AF267244.1 | - | 4 | 693 | |||
| AF267245.1 | - | 3 | 580 | |||
| AJ305370.1 | - | 6 | 966 | |||

KLRF1
Genomic Map