siRNA Details
| Gene Symbol: GALNT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017417.1 | NP_059113.1 | 3 | 677 | ||
| GenBank mRNA | AJ271385.1 | - | 3 | 677 | ||
| BC140888.1 | - | 3 | 643 | 9021756 | ||
| BC140889.1 | - | 3 | 643 | 9021757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GALNT8| Gene Symbol: GALNT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017417.1 | NP_059113.1 | 3 | 677 | ||
| GenBank mRNA | AJ271385.1 | - | 3 | 677 | ||
| BC140888.1 | - | 3 | 643 | 9021756 | ||
| BC140889.1 | - | 3 | 643 | 9021757 | ||