siRNA Details
| Gene Symbol: IFT57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018010.3 | NP_060480.1 | 11 | 1759 | ||
| GenBank mRNA | AF139576.1 | - | 11 | 1589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFT57| Gene Symbol: IFT57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018010.3 | NP_060480.1 | 11 | 1759 | ||
| GenBank mRNA | AF139576.1 | - | 11 | 1589 | ||