siRNA Details
| Gene Symbol: ITIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002216.2 | NP_002207.2 | 3 | 342 | ||
| GenBank mRNA | AK225277.1 | - | 3 | 205 | ||
| AK225329.1 | - | 3 | 247 | |||
| AK291379.1 | - | 3 | 290 | |||
| BC132685.1 | - | 3 | 229 | 40146707 | ||
| BG617651.1 | - | 3 | 343 | |||
| X07173.1 | - | 3 | 243 | |||

ITIH2
Genomic Map