siRNA Details
Gene Symbol: IL17RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018725.3 | NP_061195.2 | 2033 | |||
GenBank mRNA | BC000980.1 | - | 2003 | 2985728 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IL17RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018725.3 | NP_061195.2 | 2033 | |||
GenBank mRNA | BC000980.1 | - | 2003 | 2985728 |