siRNA Details
| Gene Symbol: CNPY3-GNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318856.1 | NP_001305785.1 | 6 | 1229 | ||
| NM_001318857.1 | NP_001305786.1 | 5 | 1101 | |||
| NM_001318858.1 | NP_001305787.1 | 5 | 993 | |||
| NR_134890.1 | 5 | 1055 | ||||
| NR_134891.1 | 4 | 958 | ||||
| NR_134892.1 | 5 | 1086 | ||||
| GenBank mRNA | BQ947041.1 | - | 6 | 862 | ||
| Gene Symbol: GNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318865.1 | NP_001305794.1 | 6 | 800 | ||
| NM_018960.5 | NP_061833.1 | 6 | 857 | |||
| NR_134899.1 | 5 | 714 | ||||
| GenBank mRNA | AF101477.1 | - | 6 | 853 | ||
| BC032627.1 | - | 6 | 849 | 5229272 | ||
| FJ224320.1 | - | 6 | 853 | |||
| KU178603.1 | - | 6 | 843 | |||
| KU178604.1 | - | 6 | 786 | |||
| KU178605.1 | - | 5 | 700 | |||

CNPY3-GNMT
Genomic Map