siRNA Details
| Gene Symbol: CHST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019886.3 | NP_063939.2 | 1 | 556 | ||
| GenBank mRNA | AB037187.1 | - | 1 | 321 | ||
| AB040711.1 | - | 1 | 324 | |||
| BC045537.1 | - | 1 | 459 | 5261787 | ||
| BQ940898.1 | - | 1 | 486 | |||

CHST7
Genomic Map