siRNA Details
| Gene Symbol: CHST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019886.3 | NP_063939.2 | 2 | 2296 | ||
| GenBank mRNA | AB037187.1 | - | 2 | 2061 | ||
| BP250418.1 | - | 1 | 404 | |||
| DA277492.1 | - | 1 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHST7| Gene Symbol: CHST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019886.3 | NP_063939.2 | 2 | 2296 | ||
| GenBank mRNA | AB037187.1 | - | 2 | 2061 | ||
| BP250418.1 | - | 1 | 404 | |||
| DA277492.1 | - | 1 | 406 | |||