siRNA Details
| Gene Symbol: KLRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114396.1 | NP_001107868.1 | 7 | 766 | ||
| NM_002262.3 | NP_002253.1 | 6 | 829 | |||
| NM_007334.2 | NP_031360.1 | 5 | 736 | |||
| XM_017019291.1 | XP_016874780.1 | 7 | 753 | |||
| GenBank mRNA | AF498040.1 | - | 6 | 647 | ||
| AF498041.1 | - | 5 | 548 | |||
| AK291506.1 | - | 7 | 771 | |||
| AY227806.1 | - | 7 | 766 | |||
| AY327501.1 | - | 5 | 634 | |||
| BC028009.1 | - | 6 | 642 | 5213081 | ||
| U30610.1 | - | 6 | 653 | |||

KLRD1
Genomic Map