siRNA Details
| Gene Symbol: CHST8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127895.1 | NP_001121367.1 | 5 | 1343 | ||
| NM_001127896.1 | NP_001121368.1 | 4 | 1254 | |||
| NM_022467.3 | NP_071912.2 | 4 | 1596 | |||
| XM_011527222.1 | XP_011525524.1 | 4 | 1577 | |||
| XM_011527224.1 | XP_011525526.1 | 5 | 1711 | |||
| XM_017027143.1 | XP_016882632.1 | 4 | 1247 | |||
| GenBank mRNA | AB047801.1 | - | 4 | 1110 | ||
| AF300612.1 | - | 4 | 1057 | |||
| AF305781.1 | - | 4 | 1153 | |||
| BC011380.1 | - | 4 | 1206 | 3874578 | ||
| BC014250.2 | - | 4 | 1250 | 4635405 | ||
| BC018723.1 | - | 4 | 1596 | 4137487 | ||

CHST8
Genomic Map