siRNA Details
| Gene Symbol: ADGRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083909.2 | NP_001077378.1 | 5 | 635 | ||
| NM_001291085.1 | NP_001278014.1 | 2 | 136 | |||
| XM_005252695.1 | XP_005252752.1 | 14 | 2458 | |||
| XM_017016779.1 | XP_016872268.1 | 4 | 495 | |||
| GenBank mRNA | AB058731.1 | - | 2 | 136 | ||
| AY181241.1 | - | 14 | 2458 | |||
| AY255564.1 | - | 1 | 19 | |||
| EF536972.1 | - | 4 | 424 | |||
More Information
Set Membership: |
|

ADGRA1
Genomic Map