siRNA Details
| Gene Symbol: ALG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033087.3 | NP_149078.1 | 2 | 1404 | ||
| NR_024532.1 | 3 | 1563 | ||||
| GenBank mRNA | AB161356.1 | - | 2 | 1361 | ||
| AK027417.1 | - | 2 | 1356 | |||
| AK027814.1 | - | 1 | 2156 | |||
| AK074704.1 | - | 2 | 1356 | |||
| AK074988.1 | - | 3 | 1541 | |||
| AK075189.1 | - | 1 | 624 | |||
| AY358697.1 | - | 2 | 1148 | |||
| BC017876.1 | - | 2 | 1354 | 4698763 | ||

ALG2
Genomic Map