siRNA Details
| Gene Symbol: IDI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033261.2 | NP_150286.1 | 5 | 819 | ||
| GenBank mRNA | AK056950.1 | - | 5 | 819 | ||
| BC017778.1 | - | 5 | 818 | 4291149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IDI2| Gene Symbol: IDI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033261.2 | NP_150286.1 | 5 | 819 | ||
| GenBank mRNA | AK056950.1 | - | 5 | 819 | ||
| BC017778.1 | - | 5 | 818 | 4291149 | ||