siRNA Details
Gene Symbol: MGAT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014275.4 | NP_055090.1 | 7 | 1102 | ||
NM_054013.3 | NP_463459.1 | 6 | 1542 | |||
XM_017008998.1 | XP_016864487.1 | 7 | 928 | |||
GenBank mRNA | AB000624.1 | - | 7 | 775 | ||
AK023137.1 | - | 6 | 1542 | |||
AK225490.1 | - | 6 | 576 | |||
AK225492.1 | - | 6 | 576 | |||
AL162067.1 | - | 4 | 209 | |||
AY358984.1 | - | 7 | 783 | |||
BC009464.1 | - | 5 | 429 | |||
BC051835.1 | - | 7 | 978 | 6196058 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145926, AK213265, AK217144, AK199018, AK195601, and others.
Panther Classification:
Gene Ontology Categories:
