siRNA Details
| Gene Symbol: B3GALT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080605.3 | NP_542172.2 | 1 | 1648 | ||
| GenBank mRNA | BG290330.1 | - | 293 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3GALT6| Gene Symbol: B3GALT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080605.3 | NP_542172.2 | 1 | 1648 | ||
| GenBank mRNA | BG290330.1 | - | 293 | |||