siRNA Details
| Gene Symbol: IFNLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170743.3 | NP_734464.1 | 5 | 634 | ||
| NM_173064.2 | NP_775087.1 | 5 | 634 | |||
| NM_173065.2 | NP_775088.1 | 5 | 634 | |||
| XM_006710394.3 | XP_006710457.2 | 4 | 698 | |||
| XM_017000479.1 | XP_016855968.1 | 4 | 663 | |||
| GenBank mRNA | AF439325.1 | - | 5 | 607 | ||
| AJ534330.1 | - | 5 | 620 | |||
| AJ534331.1 | - | 5 | 620 | |||
| AK160364.1 | - | 5 | 610 | |||
| AY129151.1 | - | 5 | 607 | |||
| AY129152.1 | - | 5 | 607 | |||
| BC101407.1 | - | 4 | 496 | |||
| BC101408.1 | - | 4 | 496 | |||
| BC101975.1 | - | 5 | 620 | |||
| BC139722.1 | - | 5 | 620 | |||
| BC140872.1 | - | 5 | 683 | 9021740 | ||
| JN591376.1 | - | 5 | 607 | |||

IFNLR1
Genomic Map