siRNA Details
Gene Symbol: GPR141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181791.1 | NP_861456.1 | 1 | 524 | ||
XM_011515374.2 | XP_011513676.1 | 7 | 1295 | |||
XM_011515375.2 | XP_011513677.1 | 6 | 1258 | |||
XM_011515383.2 | XP_011513685.1 | 2 | 624 | |||
XM_011515385.2 | XP_011513687.1 | 3 | 6475 | |||
XM_011515386.2 | XP_011513688.1 | 3 | 697 | |||
XM_011515388.2 | XP_011513690.1 | 2 | 843 | |||
XM_011515389.2 | XP_011513691.1 | 2 | 882 | |||
XM_017012160.1 | XP_016867649.1 | 5 | 791 | |||
XM_017012161.1 | XP_016867650.1 | 5 | 1045 | |||
XM_017012162.1 | XP_016867651.1 | 6 | 2094 | |||
XM_017012163.1 | XP_016867652.1 | 5 | 2058 | |||
XM_017012164.1 | XP_016867653.1 | 2 | 632 | |||
XM_017012165.1 | XP_016867654.1 | 3 | 6711 | |||
XM_017012166.1 | XP_016867655.1 | 4 | 2014 | |||
XM_017012167.1 | XP_016867656.1 | 3 | 724 | |||
GenBank mRNA | AY255538.1 | - | 1 | 323 | ||
AY288420.1 | - | 1 | 524 | |||
BC120951.2 | - | 1 | 524 | 40115152 | ||
BC120952.2 | - | 1 | 524 | 40115153 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
