siRNA Details
| Gene Symbol: OR2C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198074.4 | NP_932340.3 | 2 | 651 | ||
| GenBank mRNA | BC030717.1 | - | 2 | 649 | 4860551 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2C3| Gene Symbol: OR2C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198074.4 | NP_932340.3 | 2 | 651 | ||
| GenBank mRNA | BC030717.1 | - | 2 | 649 | 4860551 | |