siRNA Details
| Gene Symbol: ADGRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198827.3 | NP_942122.2 | 2, 3 | 663 | ||
| XM_005253566.2 | XP_005253623.1 | 2, 3 | 663 | |||
| XM_011538203.2 | XP_011536505.1 | 2, 3 | 663 | |||
| XM_011538204.2 | XP_011536506.1 | 2, 3 | 663 | |||
| XM_011538205.2 | XP_011536507.1 | 2, 3 | 663 | |||
| XM_011538206.2 | XP_011536508.1 | 2, 3 | 663 | |||
| XM_011538207.2 | XP_011536509.1 | 2, 3 | 663 | |||
| XM_011538208.2 | XP_011536510.1 | 2, 3 | 663 | |||
| XM_011538209.2 | XP_011536511.1 | 2, 3 | 663 | |||
| XM_011538210.2 | XP_011536512.1 | 2, 3 | 663 | |||
| GenBank mRNA | AY532280.1 | - | 2, 3 | 277 | ||
| BC112307.1 | - | 2, 3 | 134 | 8327775 | ||
| BC112309.1 | - | 2, 3 | 134 | 8327777 | ||
| BC143775.1 | - | 2, 3 | 134 | 9052289 | ||
| BX649116.1 | - | 2, 3 | 663 | |||
| EU523118.1 | - | 2, 3 | 178 | |||
More Information
Set Membership: |
|

ADGRD1
Genomic Map