siRNA Details
| Gene Symbol: GCDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000159.3 | NP_000150.1 | 12 | 1657 | ||
| NM_013976.3 | NP_039663.1 | 12 | 1425 | |||
| NR_102316.1 | 12 | 1712 | ||||
| NR_102317.1 | 11 | 1930 | ||||
| GenBank mRNA | AK098370.1 | - | 12 | 1625 | ||
| AK290407.1 | - | 12 | 1626 | |||
| AK296446.1 | - | 12 | 1675 | |||
| BC002579.2 | - | 12 | 1610 | 3142978 | ||
| BM855466.1 | - | 1 | 268 | |||
| U69141.1 | - | 11 | 1585 | |||

GCDH
Genomic Map