siRNA Details
| Gene Symbol: CTSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000308.3 | NP_000299.2 | 15 | 2675 | ||
| NM_001127695.2 | NP_001121167.1 | 15 | 2721 | |||
| NM_001167594.2 | NP_001161066.1 | 14 | 2624 | |||
| NR_133656.1 | 15 | 2682 | ||||
| GenBank mRNA | AB209705.2 | - | 15 | 1491 | ||
| AF086236.1 | - | 2 | 106 | |||
| AK055177.1 | - | 2 | 1807 | |||
| AK097786.1 | - | 14 | 1388 | |||
| AK172808.1 | - | 15 | 2679 | |||
| AK304536.1 | - | 14 | 1452 | |||
| AK307701.1 | - | 15 | 1464 | |||
| BC000597.2 | - | 15 | 1477 | 3346150 | ||
| BC093009.1 | - | 15 | 1484 | 30348898 | ||
| M22960.1 | - | 14 | 1445 | |||

CTSA
Genomic Map