siRNA Details
| Gene Symbol: AFG3L1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003226.1 | 11 | 1482 | |||
| NR_003227.1 | 10 | 1243 | ||||
| GenBank mRNA | AK056488.1 | - | 11 | 1480 | ||
| AK090856.1 | - | 1 | 747 | |||
| AK091293.1 | - | 11 | 1445 | |||
| AK097044.1 | - | 10 | 1257 | |||
| AK126911.1 | - | 9 | 1300 | |||
| AK128352.1 | - | 4 | 1082 | |||
| AK128804.1 | - | 5 | 636 | |||
| AL832984.1 | - | 5 | 733 | |||
| BC021245.2 | - | 9 | 1055 | |||

AFG3L1P
Genomic Map