siRNA Details
| Gene Symbol: CTSV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201575.1 | NP_001188504.1 | 2 | 121 | ||
| NM_001333.3 | NP_001324.2 | 2 | 324 | |||
| GenBank mRNA | AB001928.1 | - | 2 | 147 | ||
| AF070448.1 | - | 1 | 101 | |||
| AK310645.1 | - | 3 | 291 | |||
| AK310740.1 | - | 2 | 153 | |||
| AK312806.1 | - | 2 | 121 | |||
| AY358641.1 | - | 2 | 324 | |||
| BC023504.2 | - | 2 | 138 | 3139314 | ||
| BC110512.1 | - | 2 | 193 | 40031744 | ||
| Y14734.1 | - | 2 | 143 | |||

CTSV
Genomic Map