siRNA Details
| Gene Symbol: HGFAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297439.1 | NP_001284368.1 | 12 | 1525 | ||
| NM_001528.3 | NP_001519.1 | 11 | 1504 | |||
| GenBank mRNA | AK314099.1 | - | 12 | 1483 | ||
| BC112190.1 | - | 11 | 1504 | 8327658 | ||
| BC112192.1 | - | 11 | 1516 | 8327660 | ||
| D14012.1 | - | 11 | 1438 | |||

HGFAC
Genomic Map